Learn More
Description
- Suitable for immunohistochemistry, including paraffin-embedded and frozen tissue applications.
- Includes Bluing Reagent (30 mL), Modified Eosin Y Solution (30 mL), and Modified Mayer's Hematoxylin (30 mL).
- Features a modified Eosin Y formulation with a lower evaporation rate, improved color patterns, and enhanced surface tension.
- Produces crisp, intense blue nuclei with Modified Mayer's Hematoxylin for optimal staining contrast.
- Delivers staining results with blue nuclei, light pink cytoplasm, pink/red erythrocytes, pink collagen, and pink/rose muscle.
- Validated for use with small intestine, appendix, colon, and prostate control tissues.
- Store at 15°C to 25°C.
- Backed by a 6-month guarantee from the date of receipt.
Specifications
Specifications
| For Use With (Application) | Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Product Line | Novus |
| Quantity | 1 Kit |
| Storage Requirements | Room temperature |
| Product Type | Stain Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
