Learn More
Hematoxylin Stain Solution, Modified Harris Formulation, Mercury Free Nuclear Stain, Ricca Chemical
Shop All Ricca Chemical Company Products

Description
Includes
Components: Hematoxylin, Ethanol, Methanol, Potassium iodate, Aluminum ammonium sulfate dodecahydrate
Specifications
Specifications
| For Use With (Application) | Nuclear stain for general histology and cytology |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Product Type | Hematoxylin Stain Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.