Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemin (porcine), 97+%

Catalog No. AAA1116506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1116506 5 g
AAA1116503 1 g
AAA1116514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1116506 Supplier Thermo Scientific Chemicals Supplier No. A1116506
  • $281.50
    $224.65 / Each of 1

    Save $56.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16009-13-5 | C34H32FeN4O4 | 616.50 g/mol

Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hemin is used in cellular protection and control mechanism. It stimulates the synthesis of globulin. It is also used to study the bacterial strains and growth conditions, in the short-term colony assays and cell transfections. Further, it is used to treat the symptoms of various porphyrias. In addition, it is used in the preparation of agar medium for the bacteria culture.

Solubility
Soluble in dilute ammonia, trimethylamine, p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16009-13-5
Molecular Formula C34H32ClFeN4O4
Molecular Weight (g/mol) 651.95
MDL Number MFCD00010726
InChI Key BTIJJDXEELBZFS-HXFTUNQESA-K
Synonym chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin
PubChem CID 131675604
IUPAC Name 3-[(1Z,6Z,12Z,17Z)-5-(2-carboxyethyl)-22-chloro-15,20-diethenyl-4,10,14,19-tetramethyl-21,23,24,25-tetraaza-22-ferrahexacyclo[9.9.3.1³,⁶.1¹³,¹⁶.0⁸,²³.0¹⁸,²¹]pentacosa-1,3(25),4,6,8,10,12,14,16(24),17,19-undecaen-9-yl]propanoic acid
SMILES CC1=C(CCC(O)=O)\C2=C\C3=C(CCC(O)=O)C(C)=C4\C=C5/N=C(/C=C6\N([Fe](Cl)N34)\C(=C/C1=N2)C(C=C)=C6C)C(C=C)=C5C

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5229914
Merck Index 14,4644
Solubility Information Soluble in dilute ammonia,trimethylamine,p-toluidine and dimethylaniline. Slightly soluble in alcohol. Insoluble in water
Formula Weight 651.95
Percent Purity ≥98%
Chemical Name or Material Hemin (porcine)
Hazard Category H334
Precautionary Statement P261-P285-P304+P341-P342+P311-P501c
EINECSNumber 240-140-1
RTECSNumber LJ8080000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.