Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Hemoglobin

Catalog No. MP91004817 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP91004817 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP91004817 Supplier MP Biomedicals™ Supplier No. 091004817
Add to Cart
Add to Cart

Autoclavable preparation to be used with GC Agar Base.

From beef blood.

TRUSTED_SUSTAINABILITY

Specifications

Color Reddish-brown to maroon
Content And Storage 2°C to 8°C
Description Hemoglobin
Form Powder
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.