Learn More
Sigma Aldrich Fine Chemicals Biosciences Hemoglobin | 9008-02-0 | Unknown Quantity | Red | | | | |
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H02675MG
Encompass
Hemoglobin | Purity: N/A | Mol Wt: 64,500 Da | CAS No: 9008-02-0 | MFCD00004244 | N/AHemoglobin, the iron-containing globular protein found within red blood cells, serves as the primary carrier of oxygen from the lungs to body tissues. It makes up ~35% of red blood cell content by weight and has a heterotetrameric structure composed of 4 subunits, with each subunit containing a heme group capable of reversibly binding one oxygen molecule. The most common adult form, hemoglobin A, consists of 2 alpha and 2 beta subunits. Oxygen binding induces conformational changes in hemoglobin, altering its affinity for oxygen and allowing efficient oxygen delivery from areas of high concentration in the lungs to low concentration in respiring tissues. The iron-containing heme groups also impart the characteristic red color to blood.
Catalog No. NC2388834
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.