Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemoglobin alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674410
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-410 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-410 Supplier Invitrogen™ Supplier No. PA19527
  • $518.00
    $424.40 / Each of 1

    Save $93.60

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9527 detects Hemoglobin alpha in human samples. PA1-9527 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9527 detects a 15 kDa protein representing Hemoglobin alpha. PA1-9527 immunogen is a synthetic peptide based on the human hemoglobin alpha protein.

The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin alpha
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Hba1
Gene Accession No. P69905
Gene Alias alpha 1 globin; alpha globin 1; alpha one globin; alpha-1/2-globin; alpha-2 globin chain; alpha-globin; delta globin; GloA; globin, alpha; haemaglobin alpha 1; Hba; HBA1; HBA2; Hba-a1; Hba-a2; HBAM; Hbat1; HBA-T3; HBH; hemoglobin alpha 1 chain; hemoglobin alpha 1 globin chain; hemoglobin alpha chain; hemoglobin alpha, adult chain 1; hemoglobin alpha, adult chain 2; hemoglobin alpha-1/2 chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin subunit alpha-1/2; hemoglobin, alpha 1; LOC287167
Gene Symbols Hba1
Host Species Chicken
Immunogen Synthetic peptide mixture based on the human hemoglobin alpha protein.
Purification Method IgY Fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.