Learn More
Bioassay Systems QuantiChrom™ Hemoglobin Assay Kit. For quantitative determination of hemoglobin and evaluation of drug effects on hemoglobin metabolism.
SDP
Catalog No. 50489219 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of hemoglobin and evaluation of drug effects on hemoglobin metabolism. Key Features: Sensitive and accurate. Linear detection range 0.9 - 200 mg /dL hemoglobin in 96-well plate assay. Simple and high-throughput. The "mix-and-read" procedure involves addition of a single working reagent and reading the optical density. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Safety. Reagents do not contain any toxic components, e.g. hexacyanoferrate(III) and potassium cyanide used in Drabkin's reagent. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD400nm. Samples: Blood, serum, plasma, urine etc. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.9 mg/dL.
Catalog No. 50-489-219 Supplier Bioassay Systems Supplier No. DIHB250
For quantitative determination of hemoglobin and evaluation of drug effects on hemoglobin metabolism. Key Features: Sensitive and accurate. Linear detection range 0.9 - 200 mg /dL hemoglobin in 96-well plate assay. Simple and high-throughput. The "mix-and-read" procedure involves addition of a single working reagent and reading the optical density. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Safety. Reagents do not contain any toxic components, e.g. hexacyanoferrate(III) and potassium cyanide used in Drabkin's reagent. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD400nm. Samples: Blood, serum, plasma, urine etc. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.9 mg/dL.