Learn More
Arbor Assays Hemoglobin High Sensitivity Colorimetric Detection Kit (2 Plate)
SDP
Supplier:  Arbor Assays K013HX1
Encompass_Preferred
The DetectX Hemoglobin High Sensitivity Colorimetric Detection Kit uses a single reaction solution that is stable at 4°C, is not light sensitive, and does not contain dangerous chemicals.
Catalog No. 50-227-8396
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.