Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hemoglobin Monoclonal Antibody (HB11-231.2), Invitrogen™

Catalog No. PIMA546037 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546037 100 μg
01-671-836 500 μg
PIMA546169 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546037 Supplier Invitrogen™ Supplier No. MA546037
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10808 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10808 has been successfully used in ELISA and Western blot applications. The MA1-10808 immunogen is purified human hemoglobin.

The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hemoglobin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HB11-231.2
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HBB
Gene Accession No. P68871, P69905
Gene Alias alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1
Gene Symbols HBA1, HBB
Host Species Mouse
Immunogen Purified human hemoglobin.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3039, 3043
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.