Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Hemopexin Antibody Pair [HRP]
SDP

Catalog No. NB009657 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009657 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009657 Supplier Novus Biologicals™ Supplier No. NBP27930915PLATES

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Polyclonal/Monoclonal Antibody Pair

specifically detects in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Assay Range 18.8 pg/mL
Assay Sensitivity 18.8-1200 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human Hemopexin.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta-1B-glycoprotein, FLJ56652, hemopexin, HX
Gene ID (Entrez) 3263
Gene Symbol HPX
Host Species Rabbit/Mouse
Kit Contents
  • Detection Antibody: 0.4 mg/mL of rabbit anti-Human Hemopexin polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % glycerol, pH 7.4).
  • Capture Antibody: 1 mg/mL of mouse anti-Human Hemopexin monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 48 ng of recombinant Human Hemopexin.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
    Storage Requirements Storage is content dependent.
    Target Hemopexin
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1200 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.