Learn More
ACROBiosystems ACROBiosystems Hendra virus Fusion glycoprotein (A263T), His Tag
Supplier:  ACROBiosystems FUNH52H450UG
Encompass_Preferred
Recombinant Protein;50UG;Hendra virus Fusion glycoprotein (A263T), His Tag (FUN-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Ser 487 (Accession # O89342-1 (A263T)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.7 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Fusion Protein Antibody, Human IgG1 (5B3) at 1 μg/mL (100 μL/well) can bind Hendra virus Fusion glycoprotein (A263T), His Tag (Cat. No. FUN-H52H4) with a linear range of 1-2 ng/mL (QC tested).
Catalog No. 50-210-7188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.