Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Heneca, A2A Receptor Agonist, Solid, >99% Purity, 1 mg

Catalog No. 502707578
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7578 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7578 Supplier Abcam Supplier No. AB1204581MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective A2A receptor agonist with >99% purity.

  • Purity: >99%
  • Molecular Weight: 388.4 Da
  • Activity: Selective A2A receptor agonist (Ki = 2.2 nM) with selectivity over A1 receptors
  • Functional Effects: Displays antiaggregatory activity, increases cAMP accumulation (EC50 = 43 nM), and inhibits superoxide anion production (EC50 = 3.63 nM) in neutrophils
  • CAS Number: 141018-30-6
  • Form: Solid
  • Storage: Store at +4°C
  • Shipping: Ambient - Can Ship with Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Target Molecule A2A Receptor
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.