Learn More
Abcam Heneca, Selective A2A Receptor Agonist, 99% Purity, 10 mg
Description
- Purity: >99%
- Molecular Weight: 388.4 Da
- Biological Activity: Selective A2A receptor agonist (Ki = 2.2 nM) with good selectivity for A2A over A1 receptors.
- Functions: Displays antiaggregatory activity, increases cAMP accumulation (EC50 = 43 nM), and inhibits superoxide anion production (EC50 = 3.63 nM) in neutrophils.
- Storage: Store at +4°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | A2A Receptor |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Quantity | 10 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.