Description
- On fresh air supply for unit
- Filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 μm)
Specifications
Specifications
| Description | HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 μm)) |
| For Use With (Equipment) | BINDER Chambers FD 56 |
| Material | Plastic |
| Type | HEPA Filter |