Learn More
BINDER™ HEPA Air Filter
SDP
Catalog No. 08111730 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers FED 400
Catalog No. For Use With (Equipment)
08-111-730 BINDER Chambers FED 400
1 options
Catalog No. 08-111-730 Supplier BINDER™ Supplier No. 80120076
Add to Cart
Add to Cart
BINDER™ HEPA Air Filter is an accessory for unit FED 400 Drying and Heating Chamber.
  • On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 μm)
Order Info
ForUseWithApplication
Specifications
Description HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 μm))
For Use With (Equipment) BINDER Chambers FED 400
Type HEPA Filter