Learn More
Thermo Scientific™ Replacement HEPA Main Filter
GSA_VA
Catalog No. 15497022
Change view
Click to view available options
Type:
HEPA Filter
Catalog No. Type
15-497-022 HEPA Filter
1 options
Catalog No. 15-497-022 Supplier Thermo Scientific™ Supplier No. 760175
Add to Cart
Add to Cart
Replacement HEPA filter for use with Steri-Cycle™ Direct Heat Models, Steri-Cycle™ Water Jacket Series 2, Steri-Cycle™ Water Jacket Series 3, and Steri-Cycle™ 185 Pro CO2Incubators
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Replacement HEPA filter for use with Steri-Cycle™ Direct Heat Models, Steri-Cycle™ Water Jacket Series 2, Steri-Cycle™ Water Jacket Series 3, and Steri-Cycle™ 185 Pro CO2 Incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Replacement HEPA Filter
For Use With (Equipment) Steri-Cycle™ Direct Heat Models, Steri-Cycle™ Water Jacket Series 2, SteriCycle™ Water Jacket Series 3, and Steri-Cycle™ 185 Pro Series CO2 Incubators
Lighting LED
Type HEPA Filter