Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Antibody [DyLight 650], R&D Systems™
SDP

Catalog No. AF7276W
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7276W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF7276W Supplier R&D Systems Supplier No. AF7276W

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Sheep Polyclonal Antibody

Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 Polyclonal antibody specifically detects Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3 in Human samples. It is validated for Western Blot

Specifications

Antigen Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias h3-OST-3B, heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived human Heparan Sulfate Glucosamine 3-O-Sulfotransferase 3, Gly60-Asp390, Accession # Q9Y662
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9953
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.