Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin, Fisher BioReagents

Catalog No. BP252410 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 mg
250 mg
Packaging:
Poly Vial
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP252410 10 mg Poly Vial
BP2524250 250 mg Poly Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP252410

missing translation for 'validMainframeMsg'

Heparin is used as an anticoagulant in biochemical research.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Color White
pH 5.0 to 7.5
Loss on Drying ≤5.0%
Packaging Poly Vial
Quantity 10 mg
Merck Index 15, 4688
Vapor Pressure Negligible
Formula Weight ≈5000
Physical Form Powder or Solid
Chemical Name or Material Heparin, From Porcine Intestinal Mucosa
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.