Learn More
Apexbio Technology LLC HyperTrap Heparin HP Column(Synonyms: Heparin Affinity Column, Heparin HP Column, HyperTrap Heparin Affinity Chromatography Column), 5x5ml.
SDP
Supplier:  Apexbio Technology LLC PC100925
Encompass_Preferred
HyperTrap Heparin HP Column features the HyperChrom Heparin HP Agarose affinity chromatography resin which consists of heparin covalently immobilized on an agarose matrix Heparin is a naturally occurring glycosaminoglycan known for specific affinity interactions with various biomolecules including coagulation factors growth factors interferons lipoprotein lipase nucleic acid-binding proteins steroid receptors and antithrombin III This column enables efficient separation and purification of these proteins making it suitable for biochemical research proteomics analyses and protein characterization studies
Catalog No. 50-000-61148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.