Learn More
Specifications
Specifications
| Color | Colorless or White |
| Quantity | 100 mg |
| Formula Weight | ≈3000 |
| Physical Form | Powder or Solution |
| Chemical Name or Material | Heparin, Sodium Salt from Porcine |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15-30°C). Solutions are stable in the pH range 4-9 when stored at either room temperature or refrigerated temperatures. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.