Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin, MP Biomedicals™

Catalog No. ICN19411483 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19411483 250 mg
ICN19411410 10 mg
ICN19411450 50 mg
ICN19411480 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19411483 Supplier MP Biomedicals, Inc Supplier No. 0219411483
Add to Cart
Add to Cart

Heparin is typically used as an anticoagulant, it binds to antithrombin III, a naturally occurring plasma protease inhibitor, accelerating the rate at which antithrombin III inhibits coagulation proteases (factor Xa and thrombin)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689
Synonym Heparinic acid

Specifications

Color Colorless or White
Quantity 250 mg
Formula Weight ≈3000
Physical Form Powder or Solution
Chemical Name or Material Heparin, Sodium Salt from Porcine
Recommended Storage Store at Room Temperature (15-30°C). Solutions are stable in the pH range 4-9 when stored at either room temperature or refrigerated temperatures.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.