Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin Sodium (Laboratory), Fisher Chemical

Catalog No. H19 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100,000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H19 100,000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. H19

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The actual potency of this material will be listed on each label in USP units. 140,000 units is approximately 1g

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689

Specifications

Color White
pH 5.0 to 7.5
Packaging Glass Bottle
Quantity 100,000 Units
Vapor Pressure Negligible
Grade Laboratory
Physical Form Powder or Solid
Chemical Name or Material Heparin Sodium, Blood anticoagulant
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. The toxicological properties have not been fully investigated. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.