Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparin, Sodium Salt from Porcine, MP Biomedicals™

Catalog No. MP219411491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219411491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219411491 Supplier MP Biomedicals, Inc Supplier No. 0219411491
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heparin is a polymer classified as a mucopolysaccharide or a glycosoaminoglycan. It is biosynthesized and stored in mast cells of various mammalian tissues.

Product is made up of repeated disaccharide units consisting of 1,4-linked L-iduronic acid and D-glucosamine. The iduronic acid residues are O-sulfated at position 2, and the glucosamine residues are N-sulfated and O-sulfated at position 6. The repeated block can be interrupted or extended by residues of beta-D-glucuronic acid and 6-O-sulfated N-acetyl-alpha-D-glucosamine.

  • The amount of heparin needed to prevent coagulation in whole blood is between 20 units and 50 units per ml of whole blood
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-08-1
MDL Number MFCD00081689
Synonym Heparinic acid

Specifications

Quantity 1 g
Formula Weight about 3,000g/mol
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.