Learn More
Invitrogen™ Hepatitis B Virus Core Antigen Monoclonal Antibody (1-5)
Shop All Thermo Scientific ProductsDescription
MA1-7607 detects HBcAg from virus samples. MA1-7607 has been successfully used in Western blot, ELISA, immunoprecipitation and immunohistochemistry procedures. The MA1-7607 immunogen is a purified recombinant HBcAg core protein. The epitope corresponds to residues surrounding residue 80. Store at 4°C short term, and -80°C long term.
Specifications
Specifications
| Antigen | Hepatitis B Virus Core Antigen |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1-5 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene Alias | ayw; HBc; HBcAg; HBV; HepB |
| Host Species | Mouse |
| Immunogen | Purified recombinant HBcAg. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.