Learn More
Invitrogen™ Hepatitis B Virus Core Antigen Monoclonal Antibody (13A9)
Shop All Thermo Scientific ProductsDescription
MA1-7606 detects HBcAg from virus samples. MA1-7606 has been successfully used in ELISA, Immunoprecipitation, Western Blot and Immunohistochemistry procedures. The MA1-7606 immunogen is a purified recombinant HBcAg core protein. The epitope corresponds to residues 135-140.
Specifications
Specifications
| Antigen | Hepatitis B Virus Core Antigen |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 13A9 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | ayw; HBc; HBcAg; HBV; HepB |
| Host Species | Mouse |
| Immunogen | Purified recombinant HBcAg. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.