Learn More
Invitrogen™ Hepatitis B Virus Surface Ayw/Adw Monoclonal Antibody (HB5)

Description
This antibody recognizes the following Hepatitis B virus (HBV) subtypes: ayw2, ayw3, ayw4, ayr, adw2, adw4, adrq+, adrq-. Specificity of the antibody HB5 was verified by ELISA on panel of virus subtypes. The antibody HB5 does not cross-block with the antibody HB3 (MA1-19263).
Specifications
Specifications
| Antigen | Hepatitis B Virus Surface Ayw/Adw |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | HB5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene Alias | HBsAg; HBV; hepatitis b virus surface antigen; HepB |
| Host Species | Mouse |
| Immunogen | Purified HBsAg from human plasma. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.