Learn More
Invitrogen™ Hepatitis C Virus Core Antigen Monoclonal Antibody (11B3)
Description
MA1-7613 detects HCV from virus samples. MA1-7613 has been successfully used in ELISA, Western Blot, and Immunohistochemistry procedures. The MA1-7613 immunogen is a purified recombinant HCV core protein.
Specifications
Specifications
| Antigen | Hepatitis C Virus Core Antigen |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 11B3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | HCV core; HCVgp1; HepC; HepC core; polyprotein |
| Host Species | Mouse |
| Immunogen | Purified recombinant HCV core protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.