Learn More
Invitrogen™ Hepatitis C Virus Core Antigen NS3/NS4 Polyclonal Antibody

Description
The PA1-73104 antibody reacts with Hepatitis C virus Core, NS3, & NS4 antigens. PA1-73104 has been successfully used in ELISA and Western blot applications. The PA1-73104 antibody was raised against recombinant (E. coli) polypeptide containing core + NS3 + NS4 proteins.
Specifications
Specifications
| Antigen | Hepatitis C Virus Core Antigen NS3/NS4 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | HCV core; HepC; HepC core |
| Host Species | Goat |
| Immunogen | Recombinant (E. coli) polypeptide containing core + NS3 + NS4. |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.