Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS4b Monoclonal Antibody (2-H1)

Catalog No. 01670350
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-350 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-350 Supplier Invitrogen™ Supplier No. MA17616
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7616 detects HCV from virus samples. MA1-7616 has been successfully used in ELISA, Western blot and Immunohistochemistry procedures. The MA1-7616 immunogen is purified recombinant HCV NS4B protein, and it interacts with an epitope within residues 1710-1730. Store at 4ºC short term. For extended storage aliquot and store at -80ºC.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS4b
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2-H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias HCV; HCV NS4; Hepatitis C Virus; HepC; HepC NS4
Host Species Mouse
Immunogen Purified recombinant HCV NS4B protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.