Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis D Virus Large delta Protein Recombinant Rabbit Monoclonal Antibody (HL1051)
GREENER_CHOICE

Catalog No. PIMA542362 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542362 Supplier Invitrogen™ Supplier No. MA542362
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

HDV is the only virus in the genus, Deltaviridae. HDV is not classified into a viral family because it is a unique virus dependent on HBV. HDV is a co-infection of HBV. The envelope of HDV particles contains the Hepatitis B surface antigen (HBsAg). The production and transmission of HDV is entirely dependent on HBV to provide HBsAg. Thus, HDV is considered a satellite virus of HBV. Unlike a classical satellite virus, however, HDV does not share sequence similarity with HBV, and it can replicate independently of HBV. The nucleocapsid is made up of 60 large and small delta antigens. These are the only proteins encoded by HDV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis D Virus Large delta Protein
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1051
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias HDV
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Hepatitis D virus Large delta protein (Hepatitis delta virus genotype I (isolate D380)). The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.