Learn More
Diagnostic Biosystems HEPATOCYTE Hepatocyte Specific Antigen (OCH1E5)
Supplier:  Diagnostic Biosystems MOB42601
Encompass_Preferred
Hepatoblastoma is the most common primary tumor of the liver in children. The use of specific hepatocyte markers and also of α fetoprotein or carcinoembryonic antigen are useful for the identification of normal and malignant fetal hepatocytes. This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.
Catalog No. 50-238-5728
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.