Learn More
Bachem Hepcidin-20 (human) Trifluoroacetate
SDP
Supplier:  Bachem 4065374.1000
Encompass_Preferred
Hepcidin-20 (human) Trifluoroacetate, H-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr-OH, 342790-23-2, C85H135N27O23S9, Mr 2191.77, 1mg. Hepcidin-20, a degradation product of hepcidin-25 found in plasma and urine, shows an increased antimicrobial activity compared to the full-length peptide, but it is not involved in iron metabolism.
Catalog No. 50-194-6659
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.