Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HEPES, Free Acid, ULTROL™ Grade, Calbiochem™,

Catalog No. 3913381KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-133-81KG 1 kg
39-133-8100GM 100 g
39-133-8500GM 500 g
39-133-85KG 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 39-133-81KG Supplier MilliporeSigma™ Supplier No. 3913381KG
Add to Cart
Add to Cart

A zwitterionic N-substituted aminosulfonic acid buffer

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Quantity 1 kg
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 238.3
Grade by titration (dry basis)
Chemical Name or Material zwitterionic N-substituted aminosulfonic acid buffer
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.