Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ HepG2 Hypoxic/ Normoxic Cell Lysate
SDP

Catalog No. NBP236453
Catalog No. NBP236453 Supplier Novus Biologicals™ Supplier No. NBP236453

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

The HepG2 Hypoxic/ Normoxic Cell Lysate is derived from human. It has been validated for the following applications: SDS-Page.

Specifications

Tissue Liver
For Use With (Application) SDS-Page
Storage Buffer The protein lysate is prepared in 1X Laemmli sample buffer with BME. Boiling before loading is not necessary.
Cellular Fraction Hypoxic / Normoxic
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.