Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPPS, (Crystalline),MP Biomedicals

Catalog No. 0210192783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-101-92783 250 g
ICN10192725 25 g
02-101-92780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-101-92783 Supplier MP Biomedicals, Inc Supplier No. 0210192783
Add to Cart
Add to Cart

This is the propane analog of HEPES

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16052-06-5
Molecular Formula C9H20N2O4S
Molecular Weight (g/mol) 252.329
InChI Key OWXMKDGYPWMGEB-UHFFFAOYSA-N
Synonym hepps, epps, 4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, 1-piperazinepropanesulfonic acid, 4-2-hydroxyethyl, n-2-hydroxyethyl piperazine-n'-3-propanesulfonic acid, 4-2-hydroxyethyl piperazine-1-propanesulfonic acid, 4-2-hydroxyethyl-1-piperazinepropane sulfonic acid, 4-2-hydroxyethyl-1-piperazinepropanesulphonic acid
PubChem CID 85255
ChEBI CHEBI:42298
IUPAC Name 3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCCS(=O)(=O)O)CCO

Specifications

Physical Form Crystalline
Quantity 250 g
Formula Weight 252.3
Chemical Name or Material HEPPS
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.