Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPPSO, 99%, MP Biomedicals

Catalog No. ICN15123650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15123650 50 g
ICN15123610 10 g
ICN15123680 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15123650 Supplier MP Biomedicals, Inc Supplier No. 0215123650
Add to Cart
Add to Cart

New Zwitterionic hydrogen buffer for biological research

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-78-0
Molecular Formula C9H20N2O5S
Molecular Weight (g/mol) 268.328
InChI Key GIZQLVPDAOBAFN-UHFFFAOYSA-N
Synonym heppso, 2-hydroxy-3-4-2-hydroxyethyl piperazin-1-yl propane-1-sulfonic acid, n-hydroxyethyl piperazine-n'-2-hydroxypropanesulfonic acid, beta-hydroxy-4-2-hydroxyethyl piperazine-1-propanesulphonic acid, beta-hydroxy-4-2-hydroxyethyl-1-piperazinepropanesulfonic acid, 3-4-2-hydroxyethyl-1-piperazinyl-2-hydroxypropanesulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane-3-sulfonic acid, 4-2-hydroxyethyl piperazine-1-2-hydroxypropane sulfonic acid free acid, 2-hydroxy-3-4-2-hydroxyethyl piperazinyl propanesulfonic acid
PubChem CID 100205
ChEBI CHEBI:32951
IUPAC Name 2-hydroxy-3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
SMILES C1CN(CCN1CCO)CC(CS(=O)(=O)O)O

Specifications

Quantity 50 g
Assay Percent Range 99%
Formula Weight 268.3
Chemical Name or Material HEPPSO, Free Acid
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.