Learn More
Advanced Biomatrix Heprasil Thiol-Modified Hyaluronan/Heparin Mixture, 2 units x 5 mL
Supplier:  Advanced Biomatrix GS215F2EA
Encompass_Preferred
Heprasil is a mixture of thiol-modified hyaluronic acid thiol-modified heparin. Heprasil is a component of the HyStem-HP kits, but can be purchased separately here. Comes with 10 mL of Buffer A. HyStem V2.0 kit components are reconstituted with a buffered 1X PBS. The resulting components will also result in a pH neutral isotonic salt hydrogel. The ending pH, osmo, and rheological properties are the same from Version 1.0 to Version 2.0. Heprasil is thiol-modified hyaluronic acid with thiol-modfied heparin and is a component of the HyStem hydrogel kit. Hyaluronic acid is a major constituent of native extracellular matrix (ECM). Heparin is also present in the ECM as heparan sulfate. Most cells do not attach to Heprasil-only hydrogels. Heprasil must be used in conjunction with Gelin-S or ECM proteins such as laminin, collagen, or fibronectin for most 3-D cell culture and tissue-engineering applications.
Catalog No. NC2366528
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.