Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptachlor Epoxide (Isomer B) Solution, 1000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1164
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USEPA1164 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USEPA1164 Supplier Agilent Technologies Supplier No. EPA11641
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1024-57-3 , 67-56-1
Molecular Formula C10H5Cl7O
Molecular Weight (g/mol) 389.299
InChI Key ZXFXBSWRVIQKOD-GOWUZIGHSA-N
PubChem CID 94865622
SMILES C12C(C(C3C1O3)Cl)C4(C(=C(C2(C4(Cl)Cl)Cl)Cl)Cl)Cl

Specifications

CAS Max % 99.87, 0.13
Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Heptachlor Epoxide (Isomer B), 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.