Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptacosanoic Acid 98.0+%, TCI America™
SDP

Catalog No. H09711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09711G 1 g
H0971100MG 100 mg
H09715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H09711G Supplier TCI America Supplier No. H09711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7138-40-1
Molecular Formula C27H54O2
Molecular Weight (g/mol) 410.73
MDL Number MFCD00014029
InChI Key VXZBFBRLRNDJCS-UHFFFAOYSA-N
PubChem CID 23524
ChEBI CHEBI:78710
IUPAC Name heptacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 1 g
Formula Weight 410.73
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptacosanoic Acid
EINECSNumber (2)-0608
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.