Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptaethylene Glycol Monomethyl Ether 97.0+%, TCI America™
SDP

Catalog No. H10461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10461G 1 g
H10465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H10461G Supplier TCI America Supplier No. H10461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4437-01-8
Molecular Formula C15H32O8
Molecular Weight (g/mol) 340.41
MDL Number MFCD06201006
InChI Key AGWKUHGLWHMYTG-UHFFFAOYSA-N
Synonym mPEG7-Alcohol
PubChem CID 526555
IUPAC Name 2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 340.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptaethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.