Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluorobutyryl Chloride 95.0+%, TCI America™
SDP

Catalog No. H050825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H050825G 25 g
H05085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H050825G Supplier TCI America Supplier No. H050825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 375-16-6
Molecular Formula C4ClF7O
Molecular Weight (g/mol) 232.482
MDL Number MFCD00000707
InChI Key WFELVFKXQJYPSL-UHFFFAOYSA-N
Synonym heptafluorobutyryl chloride, perfluorobutyryl chloride, butanoyl chloride, heptafluoro, heptafluorobutanoyl chloride, heptafluorobutyric acid chloride, butanoyl chloride, 2,2,3,3,4,4,4-heptafluoro, heptafluorobutyrylchloride, acmc-209tcl, cf3cf2cf2c o cl, heptafluorobutanoyl chlorid
PubChem CID 67810
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanoyl chloride
SMILES C(=O)(C(C(C(F)(F)F)(F)F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 39°C
Quantity 25 g
UN Number 3265
Formula Weight 232.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptafluorobutyryl Chloride [for Heptafluorobutyration]
EINECSNumber (2)-3475
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.