Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptafluoropropyl Iodide (stabilized with Copper chip) 95.0+%, TCI America™
SDP

Catalog No. H059625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H059625G 25 g
H05965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H059625G Supplier TCI America Supplier No. H059625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 754-34-7
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.93
MDL Number MFCD00001061
InChI Key XTGYEAXBNRVNQU-UHFFFAOYSA-N
Synonym heptafluoro-1-iodopropane, heptafluoropropyl iodide, perfluoropropyl iodide, 1-iodoheptafluoropropane, 1-iodoperfluoropropane, propane, heptafluoro-1-iodo, 1-perfluoropropyl iodide, heptafluoroiodopropane, iodoheptafluoropropane, propane, heptafluoroiodo
PubChem CID 33977
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-3-iodopropane
SMILES FC(F)(F)C(F)(F)C(F)(F)I

Specifications

Melting Point -95°C
Color Red
Boiling Point 41°C
Quantity 25 g
Formula Weight 295.93
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptafluoropropyl Iodide (stabilized with Copper chip)
RTECSNumber TZ3930000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.