Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, ≥99.8% Solstice TraceSELECT™,

Catalog No. 60044464 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-464 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-464 Supplier Solstice Supplier No. 013791L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for inorganic trace analysis,≥99.8%

  • High purity solvents for speciation analysis by LC-ICP-MS

  • Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance

  • The blank values for metal traces in these solvents are in the ppb range or lower.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Density 0.684g/mL at 25°C
Quantity 1 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Packaging Glass Bottle
Vapor Pressure 40mmHg (20°C), 83mmHg (37.7°C)
Linear Formula CH3(CH2)5CH3
UN Number UN1206
Beilstein 1730763
Solubility Information Insoluble in water
Formula Weight 100.2g/mol
Percent Purity ≥99.8%
Grade TraceSELECT
Chemical Name or Material Heptane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331-P501
Precautionary Statement H225-H304-H315-H336-H410
DOTInformation class 3 - PG 2 - Heptanes
EINECSNumber 205-563-8
RTECSNumber MI7700000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.