Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, 99%, Reagent Grade, Solstice

Catalog No. 60048033 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-033 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-033 Supplier Solstice Supplier No. H21982.5L
Add to Cart
Add to Cart

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.5°C
Color Colorless
Density 0.684g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point −4°C (24.8°F)
Ignition Point 433°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 83mmHg (37.7°C), 40mmHg (20°C)
Linear Formula CH3(CH2)5CH3
UN Number UN1206
Beilstein 1730763
Boiling Point 98.2°C to 98.4°C (1000 hPa)
Solubility Information 0.003g/L in water at 0°C
Formula Weight 100.2g/mol
Percent Purity 0.99
Chemical Name or Material Heptane
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
DOTInformation class 3 - PG 2 - Heptanes
RTECSNumber 205-563-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.