Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, Solstice

Catalog No. 60090000 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
Packaging:
Returnable Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-000 56 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-000 Supplier Solstice Supplier No. CS20756
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC, for spectrophotometry, >99.0%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Quantity 56 L
Flash Point -4 °C
Packaging Returnable Steel Drum
Physical Form Liquid
Refractive Index 1.3872 - 1.3884
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.