Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptane, HPLC Grade, Ricca Chemical
SDP

Catalog No. RSOH000620E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOH000620E 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOH000620E Supplier Ricca Chemical Company Supplier No. RSOH000620E4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Absorbance max. 0.750, max. 0.0140
CAS Max % 100
Appearance Colorless
Quantity 20 L
Identification Pass
Packaging Metal Can
Residue after Evaporation max. 5.00 ppm
Water max. 0.02
Boiling Point 98°C
Percent Purity min. 96%
Grade HPLC
Chemical Name or Material Heptane
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.