Learn More
Description
Spectrum™ Chemical Heptane, HPLC Grade is a straight-chain alkane applied in laboratories as a non-polar mobile phase carrier solvent in high pressure liquid chromatography. All HPLC solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 200 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2 ppm |
| Water | 0.02% |
| UN Number | UN1206 |
| Boiling Point | 98.4°C |
| Percent Purity | >99% |
| Grade | High Purity |
| Chemical Name or Material | Heptane |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
