Learn More
Sigma Aldrich Fine Chemicals Biosciences Lipopolysaccharides E500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences L4005
Encompass
This product is TCA extracted from E. coli serotype O55B5. The source strain is CDC 1644-70. The LPS O55B5 has been used to stimulate human peritoneal macrophages at 1 ng/ml and to stimulate equine peritoneal macrophages at 1-100 ng/ml.
Catalog No. 50-290-7843
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.