Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptanoic Anhydride 97.0+%, TCI America™
SDP

Catalog No. H035325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H035325ML 25 mL
H0353500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H035325ML Supplier TCI America Supplier No. H035325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 626-27-7
Molecular Formula C14H26O3
Molecular Weight (g/mol) 242.359
MDL Number MFCD00009536
InChI Key DAPZDAPTZFJZTO-UHFFFAOYSA-N
Synonym heptanoic anhydride, enanthic anhydride, heptanoic acid, anhydride, heptanoyl anhydride, n-heptanoic acid anhydride, n-heptanoic anhydride, heptanoic acid, 1,1'-anhydride, oenanthic anhydride, bisenanthic anhydride, enanthic acid anhydride
PubChem CID 69376
IUPAC Name heptanoyl heptanoate
SMILES CCCCCCC(=O)OC(=O)CCCCCC

Specifications

Melting Point -12°C
Color Yellow
Boiling Point 260°C
Quantity 25 mL
UN Number 3265
Formula Weight 242.36
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Heptanoic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.