Learn More
AdipoGen Heptelidic acid,Chemical. CAS: 57710-57-3
SDP
Supplier:  AdipoGen AGCN20118C250
Encompass_Preferred
Heptelidic acid, AdipoGen Life Sciences. Chemical. CAS: 57710-57-3 (74310-84-2 deleted). Formula: C15H20O5. MW: 280.3. Isolated from Trichoderma sp. Antibiotic. Potent selective glyceraldehyde 3-phosphate dehydrogenase (GAPDH) inhibitor. Binds to the essential Cys149 residue in the catalytic site of GADPH. Anticancer compound. Selectively kills high-glycolytic cancer cells through glucose-dependent active ATP deprivation. Antimalarial. Apoptosis inhibitor. DNA fragmentation and caspase-3 activation inhibitor. Selective and competitive inhibitor of mammalian DNA polymerases beta, lambda and terminal deoxynucleotidyl transferase (TdT) in family X of pols.
Catalog No. 50-464-495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.